missing translation for 'onlineSavingsMsg'
Learn More
Learn More
m-Cresol Purple, T.S., BAKER ANALYZED™ Reagent, J.T. Baker™
High quality chemicals for laboratory and specialized industrial use
Supplier: Avantor J.T.Baker 591504
Specifications
| m-Cresol Purple | |
| BAKER ANALYZED™ Reagent | |
| 1L = 1.03kg |
| T.S. | |
| Narrow Mouth Amber Glass Bottle | |
| 100 mL |