missing translation for 'onlineSavingsMsg'
Learn More

m-Cresol Purple Sodium Salt, Reagent Grade, LabChem™

Supplier:  LabChem LC134907

Catalog No. LC134907


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

m-Cresol Purple Sodium Salt
100
Soluble in water
404.42
Amber Glass
Black/Purple to Orange
Powder
62625-31-4
C21H17O5SNa
Passes Test
Reagent
Sulfur compounds; Carbon monoxide; Carbon dioxide
10 g

Chemical Identifiers

62625-31-4
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

EINECSNumber : 263-656-9

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions