missing translation for 'onlineSavingsMsg'
Learn More

m-Cresol Purple, sodium salt, pure, water soluble

Catalog No. p-94193
Change view
Click to view available options
Quantity:
5 g
10 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC199250050 5 g Glass bottle
AC199250100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC199250050 Supplier Thermo Scientific Chemicals Supplier No. 199250050
Only null left

CAS: 62625-31-4 | C21H17NaO5S

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 62625-31-4
Molecular Formula C21H17NaO5S
MDL Number MFCD00010177
Synonym m-Cresol Purple, sodium salt

Specifications

Solubility Information Solubility in water: soluble.
Infrared Spectrum Authentic
Formula Weight 404.4
Grade Pure
Packaging Glass bottle
Visual Transition Interval from pH 7.4 (yellow) to pH 9.0 (purple)
Color Black to Brown/Orange
Quantity 5 g
Physical Form Powder
Chemical Name or Material m-Cresol Purple, sodium salt
EINECSNumber 263-656-9
TSCA TSCA

RUO – Research Use Only