missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals m-Cresol Purple, pure, indicator grade

CAS: 2303-01-7 | C21H18O5S | 382.43 g/mol

$121.01 - $186.31

Chemical Identifiers

CAS 2303-01-7
Molecular Formula C21H18O5S
Molecular Weight (g/mol) 382.43
MDL Number MFCD00005871
InChI Key OLQIKGSZDTXODA-UHFFFAOYSA-N
PubChem CID 73030
IUPAC Name 4-[3-(4-hydroxy-2-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methylphenol
SMILES CC1=C(C=CC(=C1)O)C2(C3=CC=CC=C3S(=O)(=O)O2)C4=C(C=C(C=C4)O)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC228200050
SDS View Documents
Thermo Scientific Chemicals
228200050
5 g Glass bottle
Each for $121.01
Only null left
 
AC228200100
SDS View Documents
Thermo Scientific Chemicals
228200100
10 g Glass bottle
Each for $186.31
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Chemical Identifiers

2303-01-7
382.43
OLQIKGSZDTXODA-UHFFFAOYSA-N
4-[3-(4-hydroxy-2-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methylphenol
C21H18O5S
MFCD00005871
73030
CC1=C(C=CC(=C1)O)C2(C3=CC=CC=C3S(=O)(=O)O2)C4=C(C=C(C=C4)O)C

Specifications

2303-01-7
MFCD00005871
Authentic
4-[3-(4-hydroxy-2-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methylphenol
73030
Pure
900 min. (578nm, in buffer)
Green
Fine Crystalline Powder
C21H18O5S
OLQIKGSZDTXODA-UHFFFAOYSA-N
CC1=C(C=CC(=C1)O)C2(C3=CC=CC=C3S(=O)(=O)O2)C4=C(C=C(C=C4)O)C
382.43
382.42
Glass bottle
from pH 7.4 (yellow) to pH 9.0 (purple)
5 g
m-Cresol Purple
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 218-960-6

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only