Learn More
m-Cresol, 99%
CAS: 108-39-4 | C7H8O | 108.14 g/mol
$68.01 - $312.93
Chemical Identifiers
| CAS | 108-39-4 |
|---|---|
| Molecular Formula | C7H8O |
| Molecular Weight (g/mol) | 108.14 |
| MDL Number | MFCD00002302 |
| InChI Key | RLSSMJSEOOYNOY-UHFFFAOYSA-N |
| Synonym | 3-Methylphenol |
| PubChem CID | 342 |
| ChEBI | CHEBI:17231 |
| IUPAC Name | 3-methylphenol |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AAA1073330
|
N/A | 250 g |
Each for $68.01
|
|
|||||
|
AAA107330B
|
N/A | 1000 g |
Each for $167.48
|
|
|||||
|
AAA107330E
|
N/A | 2500 g |
Each for $312.93
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Applicationsm-Cresol is employed in the preparation of synthetic resins like phenol-formaldehyde resins, disinfectants, and photographic developers. It is used as industrial solvent for dissolving polymers such as polyaniline. It acts as a precursor to vitamin E and antiseptic amylmetacresol. It is used as insuline preservatives.
Solubility
Miscible with water, diethylether, chloroform and ethanol.
Notes
Hygroscopic. Air and light sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, strong bases, strong acids, aluminum and aluminum alloys.
Chemical Identifiers
| C7H8O | |
| MFCD00002302 | |
| 3-Methylphenol | |
| CHEBI:17231 |
Specifications
| 11°C to 12°C | |
| 108-39-4 | |
| MFCD00002302 | |
| 506719 | |
| 3-Methylphenol | |
| RLSSMJSEOOYNOY-UHFFFAOYSA-N | |
| 3-methylphenol | |
| 342 | |
| 108.14 | |
| Phenol-like | |
| 1.034 | |
| 86°C (186°F) |
| 202°C to 204°C | |
| C7H8O | |
| UN2076 | |
| 14,2579 | |
| Miscible with water,diethylether,chloroform and ethanol. | |
| CC1=CC(=CC=C1)O | |
| 108.14 | |
| CHEBI:17231 | |
| 99% | |
| Light sensitive | |
| 5 | |
| m-Cresol |
Safety and Handling
GHS H Statement
H301-H311-H314-H318-H227
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H227-H301+H311-H314-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CRESOLS, LIQUID
EINECSNumber : 203-577-9
RTECSNumber : GO6125000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only