missing translation for 'onlineSavingsMsg'
Learn More

m-Anisoyl chloride, 99%

Catalog No. p-3735529
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC212970050 5 g Glass bottle
AC212970250 25 g Glass bottle
AC212971000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC212970050 Supplier Thermo Scientific Chemicals Supplier No. 212970050
Only null left

CAS: 1711-05-3 | C8H7ClO2 | 170.6 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1711-05-3
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.6
MDL Number MFCD00000673
InChI Key RUQIUASLAXJZIE-UHFFFAOYSA-N
Synonym m-anisoyl chloride, benzoyl chloride, 3-methoxy, m-methoxybenzoyl chloride, 3-anisoyl chloride, m-anisic acid chloride, 3-methoxy-benzoyl chloride, benzoyl chloride, m-methoxy, 3-methoxybenzoic acid chloride, m-anisoylchloride, 3-anisoylchlorid
PubChem CID 74374
IUPAC Name 3-methoxybenzoyl chloride
SMILES COC1=CC=CC(=C1)C(=O)Cl

Specifications

Color Colorless to Yellow
Density 1.2140g/mL
Boiling Point 123°C to 125°C (15.0 mmHg)
Flash Point 92°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.5575 to 1.5595
Linear Formula CH3OC6H4COCl
Quantity 5 g
Beilstein 10, 140
Specific Gravity 1.214
Solubility Information Solubility in water: decomposes
Formula Weight 170.6
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material m-Anisoyl chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 216-975-2

RUO – Research Use Only