missing translation for 'onlineSavingsMsg'
Learn More

Gilson™ M-96 Well Temperature Control Plate Adapter

Catalog No. F1077604
Change view
Click to view available options
For Use With (Equipment):
PLATEMASTER™, PIPETMAX™
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
F1077604 PLATEMASTER™, PIPETMAX™
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F1077604 Supplier Gilson™ Supplier No. F1077604

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The M-96 Well Temperature Control Plate Adapter is a 96-well adapter providing passive temperature control to keep your samples hot or cold.

The Alu-heater Block is an adapter for PLATEMASTERTM and PIPETMAXTM that accommodates non-skirted and semi-skirted plates.

  • Provides passive temperature control (hot or cold) for your samples
  • 96-wells for holding samples

Specifications

Product Type Pipetter Accessory
Autoclavable Not Autoclavable
Certifications/Compliance ISO 9001
Color Gray
Device Holds 96 Tubes
Material Polypropylene
Volume (Metric) 0.2 mL
For Use With (Application) Liquid Handling, Temperature Control of Samples
For Use With (Equipment) PLATEMASTER™, PIPETMAX™
Quantity 1
Volume (English) 0.007 oz.
Warranty 1 Year
Width (English) .04 in
Height (English) .11 in
Height (Metric) 3 kg
Weight (English) .02 lb
Weight (Metric) .01 kg
No. per Pack 1
Show More Show Less