Learn More
Biotium LysoView™ 633, 100uL
Catalog No. 501964766
Encompass
LysoView™ dyes are fluorescent stains for imaging lysosome localization and morphology in live cells. The dyes accumulate in the low pH environment of the lysosomes, resulting in highly specific lysosomal staining without the need for a wash step. Far-red fluorescent LysoView™ 633 exhibits pH-sensitive fluorescence (Ex/Em 634/659 nm at pH 5), further enhancing the specificity of staining.
Catalog No. 501964766 Supplier Biotium Supplier No. 70058T
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
LysoView™ dyes are fluorescent stains for imaging lysosome localization and morphology in live cells. The dyes accumulate in the low pH environment of the lysosomes, resulting in highly specific lysosomal staining without the need for a wash step. Far-red fluorescent LysoView™ 633 exhibits pH-sensitive fluorescence (Ex/Em 634/659 nm at pH 5), further enhancing the specificity of staining.