Lysophosphatidic Acid (LPA) ELISA Kit for detecting LPA in serum, plasma, other biological fluids. Species Reactivity: Universal. Detection Range: 123.5-10,000ng/ml. Sensitivity: 50.5ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Target Synonyms: LysoPA; 1-Oleoyl-sn-Glycero-3-Phosphate; Lysophosphatidyl Acid. Restrictions: For Research Use Only. Not for use in diagnostic procedures.