missing translation for 'onlineSavingsMsg'
Learn More

Lycopodium Powder, Reagents

Catalog No. p-200020898
Change view
Click to view available options
Quantity:
25 g
100 g
Catalog No. Quantity
01337194 100 g
01337193 25 g
2 options

Catalog No. 01337194

Supplier: Labchem Inc C2236000100C2

Only null left

Lycopodium Powder, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 8023-70-9
Synonym Club moss spores

Specifications

Color Yellowish
Packaging Glass Bottle
Quantity 100 g
Specific Gravity 0.26
Grade Reagent
Physical Form Solid
Chemical Name or Material Lycopodium Powder