missing translation for 'onlineSavingsMsg'
Learn More

Lycopene, >90%, MP Biomedicals™

Catalog No. p-4405874
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19868401 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19868401 Supplier MP Biomedicals Inc Supplier No. 0219868401

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 502-65-8
Molecular Formula C40H56
Molecular Weight (g/mol) 536.888
InChI Key OAIJSZIZWZSQBC-GYZMGTAESA-N
Synonym lycopene, psi,psi-carotene, all-trans-lycopene, trans-lycopene, lycopene 7, lycored, lycopene van, lycopene, all-trans, unii-sb0n2n0wv6, ccris 7925
PubChem CID 446925
ChEBI CHEBI:15948
IUPAC Name (6E,8E,10E,12E,14E,16E,18E,20E,22E,24E,26E)-2,6,10,14,19,23,27,31-octamethyldotriaconta-2,6,8,10,12,14,16,18,20,22,24,26,30-tridecaene
SMILES CC(=CCCC(=CC=CC(=CC=CC(=CC=CC=C(C)C=CC=C(C)C=CC=C(C)CCC=C(C)C)C)C)C)C

Specifications

Color Red
Quantity 1 mg
Formula Weight 536.9
Percent Purity >90%
Physical Form Crystalline Needles
Chemical Name or Material Lycopene