missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LY294002
Description
LY294002 is a potent and specific cell-permeable inhibitor of phosphatidylinositol 3-kinase (PI3-kinase).
Format/Formulation: Lyophilized
Specifications
Specifications
| Content And Storage | Store in freezer (-5 to -30°C). |
| Gene ID (Entrez) | 5294 |
| Purification Method | TLC |
| Shipping Condition | Dry Ice |
| Specific Inhibit or Target | PI3-Kinase |
| Sterility | Non-sterile |
| Quantity | 5 mg |
| Product Type | Inhibitor |
| Inhibitor Properties | Chemical |
| Molecular Weight (g/mol) | 307.4 |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.