Learn More
Ametek LV SPINDLE
Catalog No. NC9018602
Encompass
Spindle, Entry Code: 63, SMC: 128, SRC: 0, Entry Code Allows DV-II+Pro to Calculate Viscosity, Shear Rate and Shear Stress Values, SMC Used for Viscosity and Shear Stress Calculations, SRC Used for Shear Rate and Shear Stress Calculations
Catalog No. NC9018602 Supplier Ametek Supplier No. LV3
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Spindle, Entry Code: 63, SMC: 128, SRC: 0, Entry Code Allows DV-II+Pro to Calculate Viscosity, Shear Rate and Shear Stress Values, SMC Used for Viscosity and Shear Stress Calculations, SRC Used for Shear Rate and Shear Stress Calculations