Learn More
Ametek Spindle, LV-1 (61) 15-20K, F/ Brookfield Viscometers/Rheometers
Catalog No. NC0312531
Encompass
All standard LV torque Brookfield Viscometers/Rheometers are supplied with a standard spindle set (LV-1 through LV-4) constructed of #302 stainless steel. Additional spindle options are available in #316 stainless steel or Teflon coating for increased corrosion resistance. An optional spindle rack makes a convenient storage "bench" and allows easy and quick access to all spindles.
Catalog No. NC0312531 Supplier Ametek Supplier No. LV1
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
All standard LV torque Brookfield Viscometers/Rheometers are supplied with a standard spindle set (LV-1 through LV-4) constructed of #302 stainless steel. Additional spindle options are available in #316 stainless steel or Teflon coating for increased corrosion resistance. An optional spindle rack makes a convenient storage "bench" and allows easy and quick access to all spindles.