missing translation for 'onlineSavingsMsg'
Learn More

Luzindole 98.0+%, TCI America™

Catalog No. L031625MG
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L031625MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. L031625MG Supplier TCI America Supplier No. L031625MG

Chemical Identifiers

CAS 117946-91-5
Molecular Formula C19H20N2O
Molecular Weight (g/mol) 292.38
MDL Number MFCD00672498
InChI Key WVVXBPKOIZGVNS-UHFFFAOYSA-N
Synonym luzindole, n-acetyl-2-benzyltryptamine, 2-benzyl-n-acetyltryptamine, n-2-2-benzyl-1h-indol-3-yl ethyl acetamide, n-2-2-phenylmethyl-1h-indol-3-yl ethyl acetamide, tocris-0877, n-acetyl-2-benzyl-tryptamine, acetamide, n-2-2-phenylmethyl-1h-indol-3-yl ethyl, n-2-2-benzylindol-3-yl ethyl acetamide
PubChem CID 122162
IUPAC Name N-[2-(2-benzyl-1H-indol-3-yl)ethyl]acetamide
SMILES CC(=O)NCCC1=C(CC2=CC=CC=C2)NC2=CC=CC=C12

Specifications

Melting Point 106°C
Color White-Yellow
Quantity 25 mg
Formula Weight 292.38
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Luzindole
RTECSNumber AC7765700
TSCA No
Recommended Storage Freezer (-20 °C)