missing translation for 'onlineSavingsMsg'
Learn More

Luzindole, 97%, Thermo Scientific Chemicals

Catalog No. AAJ61915LB0
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61915LB0 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ61915LB0 Supplier Thermo Scientific Chemicals Supplier No. J61915LB0

A melatonin receptor antagonist

Luzindole significantly antagonizes melatonin-mediated antinociception, suggesting that MEL-1B-R plays a further role as a mediator of melatonin modulation of pain signaling. It is used both in vitro and in vivo to evaluate the roles of melatonin receptor signaling in diverse pathways, including circadian rhythms, animal behavior, and melanophore response

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Luzindole significantly antagonizes melatonin-mediated antinociception, suggesting that MEL-1B-R plays a further role as a mediator of melatonin modulation of pain signaling. It is used both in vitro and in vivo to evaluate the roles of melatonin receptor signaling in diverse pathways, including circadian rhythms, animal behavior, and melanophore response

Solubility
Soluble in DMSO (100 mM), ethanol (100 mM), chloroform, methanol, and water.

Notes
Protect from heat. Incompatible with heat and oxidizing agents. Store at 4°C.

Chemical Identifiers

CAS 117946-91-5
Molecular Formula C19H20N2O
Molecular Weight (g/mol) 292.38
MDL Number MFCD00672498
InChI Key WVVXBPKOIZGVNS-UHFFFAOYSA-N
Synonym luzindole, n-acetyl-2-benzyltryptamine, 2-benzyl-n-acetyltryptamine, n-2-2-benzyl-1h-indol-3-yl ethyl acetamide, n-2-2-phenylmethyl-1h-indol-3-yl ethyl acetamide, tocris-0877, n-acetyl-2-benzyl-tryptamine, acetamide, n-2-2-phenylmethyl-1h-indol-3-yl ethyl, n-2-2-benzylindol-3-yl ethyl acetamide
PubChem CID 122162
IUPAC Name N-[2-(2-benzyl-1H-indol-3-yl)ethyl]acetamide
SMILES CC(=O)NCCC1=C(CC2=CC=CC=C2)NC2=CC=CC=C12

Specifications

Melting Point 44°C to 46°C
Color White to Brown
Quantity 5 mg
Solubility Information Soluble in DMSO (100 mM),ethanol (100 mM),chloroform,methanol,and water.
Formula Weight 292.37
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Luzindole
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only