missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Luxol™ Fast Blue MBSN, pure

CAS: 1328-51-4 | C32H14CuN8Na2O6S2 | 780.16 g/mol

Supplier:  Thermo Scientific Chemicals 212170250

 View more versions of this product

Catalog No. AC212170250


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Luxol™ Fast Blue MBSN
1328-51-4
MFCD00071424
Solubility in water: insoluble. Other solubilities: soluble in acohols like methanol, ethanol, propano, ls; very soluble in gylcols, ethanolamine, choline, . insoluble in hydrocarbons.
Authentic
780.16
778.15
Glass bottle
Blue
Powder
Pure
C32H14CuN8Na2O6S2
Solvent Blue 38
DKBXPLYSDKSFEQ-UHFFFAOYSA-L
C1=CC=C2C(=C1)C3=NC4=C5C=CC=CC5=C([N-]4)N=C6C7=C(C=CC=C7S(=O)(=O)[O-])C(=N6)N=C8C9=C(C(=CC=C9)S(=O)(=O)[O-])C(=N8)N=C2[N-]3.[Na+].[Na+].[Cu+2]
92030797
Pure
Complies
25 g

Chemical Identifiers

1328-51-4
780.16
DKBXPLYSDKSFEQ-UHFFFAOYSA-L
92030797
C32H14CuN8Na2O6S2
MFCD00071424
Solvent Blue 38
C1=CC=C2C(=C1)C3=NC4=C5C=CC=CC5=C([N-]4)N=C6C7=C(C=CC=C7S(=O)(=O)[O-])C(=N6)N=C8C9=C(C(=CC=C9)S(=O)(=O)[O-])C(=N8)N=C2[N-]3.[Na+].[Na+].[Cu+2]
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only