missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lutetium pieces, distilled dendritic, 99.9% (REO)
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under argon
Specifications
Specifications
| Color | Silver |
| Quantity | 1 g |
| Odor | Odorless |
| Form | Pieces |
| UN Number | UN3178 |
| Percent Purity | 99.9% |
| Assay | (REO) |
| Chemical Name or Material | Lutetium pieces, Distilled dendritic |
Safety and Handling
| Hazard Category | H228 |
| Hazard Statement | GHS H Statement H228-H260 Flammable solid. In contact with water releases flammable gases which may ignite spontaneously. |
| Precautionary Statement | P210-P240-P241-P280-P370+P378q |
| DOTInformation | Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S. |
| EINECSNumber | 231-103-0 |
| TSCA | Yes |
| storageNote1 |
|
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only