Learn More
Lutetium(III) sulfate octahydrate, 99.9% (REO), Thermo Scientific Chemicals
$307.16 - $307.16
Description
Lutetium Sulfate also called Lutetium Sulphate, is applied in making phosphors. Stable Lutetium can be used as catalysts in petroleum cracking in refineries and can also be used in alkylation, hydrogenation, and polymerization applications. It can also be used as an ideal host for X-ray phosphors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 13473-77-3 | |
| MFCD00011499 |
Specifications
| 13473-77-3 | |
| 1 g | |
| 14,5615 | |
| 782.24 (683.12 Anhydrous) | |
| Hygroscopic | |
| (REO) |
| Crystalline | |
| MFCD00011499 | |
| Soluble in water,moderately soluble in strong mineral acids. | |
| 99.9% | |
| Lutetium(III) sulfate octahydrate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only