missing translation for 'onlineSavingsMsg'
Learn More

Lutetium(III) fluoride, REacton™, 99.9% (REO), Thermo Scientific Chemicals

$635.51 - $635.51

Chemical Identifiers

CAS 13760-81-1
MDL Number MFCD00016191
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1462006
View Documents
Thermo Scientific Chemicals
01462006
5 g
Each for $635.51
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13760-81-1
MFCD00016191

Specifications

1182°C
Lump
MFCD00016191
231.97
Reacton
Hygroscopic
Lutetium(III) fluoride
13760-81-1
5 g
UN3288
99.9%
Odorless
8.3 g/mL
(REO)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.

EINECSNumber : 237-355-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only