missing translation for 'onlineSavingsMsg'
Learn More

Lutetium foil, 0.1mm (0.004in) thick, 99.9% (REO), Thermo Scientific Chemicals

Catalog No. p-7048348
Change view
Click to view available options
Quantity:
25 x 25 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA10228FF 25 x 25 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA10228FF Supplier Thermo Scientific Chemicals Supplier No. 10228FF
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon

Chemical Identifiers

CAS 7439-94-3
Molecular Formula Lu
Molecular Weight (g/mol) 174.97
MDL Number MFCD00011098
InChI Key OHSVLFRHMCKCQY-UHFFFAOYSA-N
Synonym lutecium, unii-5h0doz21uj, 5h0doz21uj, foil, foil, 0.127mm 0.005in thick reo, cassiopium, lutecio, cassiopeium, atom, pieces
PubChem CID 23929
ChEBI CHEBI:33382
IUPAC Name lutetium
SMILES [Lu]

Specifications

Color Silver to Gray
Quantity 25 x 25 mm
Odor Odorless
Packaging Mylar under Argon
Weight ≈0.62g/25x25mm
Form Foil
UN Number UN3178
Percent Purity 99.9%
Assay (REO)
Chemical Name or Material Lutetium foil, 0.1mm (0.004 in.) thick
Hazard Category H228
Hazard Statement GHS H Statement
H260-H228
In contact with water releases flammable gases which may ignite spontaneously.
Flammable solid.
Precautionary Statement P210-P240-P241-P280-P370+P378q
DOTInformation Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S.
EINECSNumber 231-103-0
TSCA Yes
storageNote1
  • Packaged under argon
Recommended Storage Ambient temperatures

RUO – Research Use Only