missing translation for 'onlineSavingsMsg'
Learn More

Lutein, 90%

Catalog No. AC456160050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC456160050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC456160050 Supplier Thermo Scientific Chemicals Supplier No. 456160050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 127-40-2 | C40H56O2 | 568.87 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 127-40-2
Molecular Formula C40H56O2
Molecular Weight (g/mol) 568.87
MDL Number MFCD08435941
InChI Key KBPHJBAIARWVSC-HZNSELMYSA-N
Synonym lutein
PubChem CID 102514741
IUPAC Name (1S)-4-[(1E,3Z,5E,7Z,9E,11Z,13E,15Z,17E)-18-[(1R)-4-hydroxy-2,6,6-trimethylcyclohex-2-en-1-yl]-3,7,12,16-tetramethyloctadeca-1,3,5,7,9,11,13,15,17-nonaenyl]-3,5,5-trimethylcyclohex-3-en-1-ol
SMILES CC1=C(C(CC(C1)O)(C)C)C=CC(=CC=CC(=CC=CC=C(C)C=CC=C(C)C=CC2C(=CC(CC2(C)C)O)C)C)C

Specifications

Color Red
Assay Percent Range 88% min. (HPLC)
Quantity 5 g
Formula Weight 568.87
Percent Purity 90%
Physical Form Powder
Chemical Name or Material Lutein
EINECSNumber 204-840-

RUO – Research Use Only