missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Luperox(R) P, tert-Butyl peroxybenzoate

Catalog No. 159042500G
Encompass
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
159042500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 159042500G Supplier Sigma Aldrich Supplier No. 159042500G

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Tert-butyl peroxybenzoate (tbpb)-mediated 2-isocyanobiaryl insertion with 1,4-dioxane has been reported.luperox was employed as polymerization and cross-linking catalyst. it was also was employed as initiator during:
  • Grafting of 2,2,6,6-tetramethyl-1-piperidinyloxy (tempo)-4-oxyacetamido-(3 propyltriethoxysilane) to poly(ethylene co-octene)
  • Preparation of conformal poly(cyclohexyl methacrylate) thin films via initiated chemical vapor depositionproduct of arkema inc

Specifications

CAS 614-45-9
Density 1.021 g/mL (at 25°C (literature))
Boiling Point 75°C to 76°C (0.2 mmHg)
Molecular Formula C11H14O3
Refractive Index n20/D 1.499 (literature)
Linear Formula C6H5COOOC(CH3)3
MDL Number MFCD00008802
Quantity 500 g
Synonym tert-Butyl peroxybenzoate; tert-Butyl perbenzoate
Molecular Weight (g/mol) 194.23
Percent Purity 98%
Show More Show Less