Learn More
New England Biolab Ltd Luna Universal Probe One-Step RT-qPCR Kit 2500 reactions
Catalog No. 501526346
Encompass
In the Luna Universal One-Step Probe RT-qPCR Kit, Hot Start Taq DNA Polymerase is combined with a novel WarmStart-activated reverse transcriptase, allowing dual control of enzyme activity via reversible, aptamer-based inhibition. This temperature-dependent activation helps to prevent undesirable non-specific priming and extension prior to thermocycling, providing added security for setting up reactions at room temperature. The engineered WarmStart Luna Reverse Transcriptase also possesses higher thermostability than many other RTs, allowing an optimal reaction temperature of 55C. For difficult targets/templates, higher RT step temperatures of up to 60C can be used without compromising Luna performance. Note that to ensure full activation of the WarmStart Luna RT, incubation at temperatures lower than 50C is not recommended.
Catalog No. 501526346 Supplier New England Biolab Ltd Supplier No. E3006E
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
In the Luna Universal One-Step Probe RT-qPCR Kit, Hot Start Taq DNA Polymerase is combined with a novel WarmStart-activated reverse transcriptase, allowing dual control of enzyme activity via reversible, aptamer-based inhibition. This temperature-dependent activation helps to prevent undesirable non-specific priming and extension prior to thermocycling, providing added security for setting up reactions at room temperature. The engineered WarmStart Luna Reverse Transcriptase also possesses higher thermostability than many other RTs, allowing an optimal reaction temperature of 55C. For difficult targets/templates, higher RT step temperatures of up to 60C can be used without compromising Luna performance. Note that to ensure full activation of the WarmStart Luna RT, incubation at temperatures lower than 50C is not recommended.