Learn More
New England Biolab Ltd Luna Universal One-Step RT-qPCR Kit 500 reactions
Numéro de catalogue. 501526344
Encompass
In the Luna One-Step RT-qPCR Kit, Hot StartTaqDNA Polymerase is combined with a novel WarmStart-activated reverse transcriptase, allowing dual control of enzyme activity via reversible, aptamer-based inhibition. This temperature-dependent activation helps to prevent undesirable non-specific priming and extension prior to thermocycling, providing added security for setting up reactions at room temperature. The engineered WarmStart Luna Reverse Transcriptase also possesses higher thermostability than many other RTs, allowing an optimal reaction temperature of 55C. For difficult targets/templates, higher RT step temperatures of up to 60C can be used without compromising Luna performance. Note that to ensure full activation of the WarmStart Luna RT, incubation at temperatures lower than 50C is not recommended.
Numéro de catalogue. 501526344 Fournisseur New England Biolab Ltd Code fournisseur E3005L
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
In the Luna One-Step RT-qPCR Kit, Hot StartTaqDNA Polymerase is combined with a novel WarmStart-activated reverse transcriptase, allowing dual control of enzyme activity via reversible, aptamer-based inhibition. This temperature-dependent activation helps to prevent undesirable non-specific priming and extension prior to thermocycling, providing added security for setting up reactions at room temperature. The engineered WarmStart Luna Reverse Transcriptase also possesses higher thermostability than many other RTs, allowing an optimal reaction temperature of 55C. For difficult targets/templates, higher RT step temperatures of up to 60C can be used without compromising Luna performance. Note that to ensure full activation of the WarmStart Luna RT, incubation at temperatures lower than 50C is not recommended.