missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One LUMITRAC™ 200 μClear™ Bottom Medium-Binding 96-well Microplate
GREENER_CHOICE

Catalog No. 07000097
Change view
Click to view available options
Color:
White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
07000097 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07000097 Supplier Greiner Bio-One Supplier No. 655095
Only null left

Free of detectable DNase, RNase, human DNA and pyrogens

Specifications

Format Standard
Lid Without Lid
Detection Method Luminescence
Array 8 x 12
Autoclavable Not Autoclavable
Binding Type Medium Binding
Color White
Material Polystyrene
No. of Wells 96
Volume (Metric) Working 25 to 340 μL
Sterility Non-sterile
Total Volume (Metric) Well 392 μL
Well Shape Round, Flat Bottom (F-well)
Dnase Free Yes
Max. RCF 4800 x g (swinging bucket rotor)
No. per Case 40
No. per Pack 10
Quantity 40/Cs.
RNase Free Yes
Well Color White
Well Design Chimney
Dimensions (L x W x H) 127.76 x 85.48 x 14.40 mm
Show More Show Less