Learn More
Research Products Corporation Luminox Cleaner, 1 Gallon
Catalog No. 50212166
Encompass
Low foaming, phosphate-free, neutral pH detergent for use in manual, ultrasonic, and machine cleaning. Safe-handling detergent used for non-corrosive cleaning without chelation or alkalinity waste treatment problems. Recommended for aluminum and soft metal surface preparation, medical devices, pharmaceutical process equipment, alkaline sensitive lab and glassware, and medical instruments in aluminum trays.
Catalog No. 50212166 Supplier Research Products Corporation Supplier No. 114013
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Low foaming, phosphate-free, neutral pH detergent for use in manual, ultrasonic, and machine cleaning. Safe-handling detergent used for non-corrosive cleaning without chelation or alkalinity waste treatment problems. Recommended for aluminum and soft metal surface preparation, medical devices, pharmaceutical process equipment, alkaline sensitive lab and glassware, and medical instruments in aluminum trays.