Learn More
Invitrogen™ Luminex™ xMAP™ INTELLIFLEX System
The Luminex xMAP INTELLIFLEX System is the newest addition to the xMAP line of instruments and is available in two configurations: the Luminex xMAP INTELLIFLEX System and the Luminex xMAP INTELLIFLEX DR-SE System.
Supplier: Invitrogen™ APX2020
Description
The Luminex xMAP INTELLIFLEX System is the newest addition to the xMAP line of instruments. It comes in two configurations: the Luminex xMAP INTELLIFLEX System (Cat. No. APX2020) and the Luminex xMAP INTELLIFLEX DR-SE System (Cat. No. APX2021). The main differences between the configurations are the dual reporter and side eject features available in the DR-SE configuration.
The Luminex xMAP INTELLIFLEX System represents the most advanced and versatile multiplexing platform, featuring an embedded PC with a touchscreen interface and a reduced footprint compared to the Luminex 200 and FLEXMAP 3D systems. With automated start up, shut down, and maintenance routines, the Luminex xMAP INTELLIFLEX System is easy to use and maintain, providing fast read times for both 96- and 384-well plate formats. The system is compatible with all Invitrogen ProcartaPlex multiplex immunoassays and Invitrogen QuantiGene Plex multiplex gene expression assays.
Features of the Luminex xMAP INTELLIFLEX System include:
- Reporter laser (RP1): 532-nm diode-pumped solid-state laser (DPSS); mode of operation, continuous wave (CW); output power varies based on mode with maximum output power of 50 mW
- Reporter detector (RP1): photomultiplier tube, detection bandwidth of 565 to 585 nm
- Dynamic range (typical): RP1: ≥5.5 logs
- Read time: 96-well plate in ∼20 min (up to 144,000 tests/hour); 384-well plate in ∼75 min (up to 153,600 tests/hour)
- System warmup time: 30 min.
- System intialization: ≤45 min.
- System verification: 5 min.
- Multiplexing: up to 500 analytes in a single sample (assay-dependent)
- Available options: service contracts, IQ/OQ
- Integrated PC and integrated barcode reader
Luminex xMAP technology
The open architecture of Luminex xMAP technology uses microspheres, digital signal processing, and traditional chemistry—combining proven technologies in a unique way.
Advantages of Luminex xMAP technology include:
- Helps reduce costs and labor through multiplexing
- Smaller sample size requirements compared to single result assays
- Enables fast, reproducible results from favorable kinetics of liquid bead array approach
- Broad coverage of applications, including protein expression and gene expression profiling
How Luminex xMAP technology works
Sets of magnetic microspheres are color-coded with combinations of red and infrared fluorophores. Each bead set is coated with a reagent specific to a particular bioassay, allowing the capture and detection of several specific analytes from a single sample. Optics in the compact analyzer excite the internal dyes that identify each microsphere as well as any reporter dye captured, providing quantitation of individual analytes on each microsphere. The analyzer reads many beads from each set, enabling rapid and precise results for several targets within a single sample.
Specifications
| • Luminex xMAP INTELLIFLEX instrument • xMAP INTELLIFLEX Software (installed and USB) • xMAP INTELLIFLEX Quick Guide • Power cable, country specific • xMAP Sheath Fluid PLUS, 20 L • Sample probe needles (3) • Off plate reagent Block • Heater block for 96-well microtiter plates • Heater block for 384-well microtiter plates • Empty Cubitainer for waste • Integrated PC with Windows 10 and integrated barcode reader • Sample Probe Height Adjustment Kit (shipped separately) • INTELLIFLEX Calibration Kit • INTELLIFLEX Performance Verification Kit |
|
| Reporter Laser (532 nm) | |
| Room Temperature |
| 96- or 384-well Plate | |
| Luminex™, xMAP™ | |
| 1 system |
For Research Use Only. Not for use in diagnostic procedures.