Learn More
Electron Microscopy Sciences LUMINEX CLEANING CLOTH
Supplier:  Electron Microscopy Sciences 7171612
Encompass
For superior cleaning of delicate surfaces. Based on advanced microfiber technology, the Dust-Off/Luminex Microfiber Cleaning Cloth is a superior cleaning product. It actually gets under dirt and oily films and lifts them away, leaving the surface clean, polished and lint and smear free. Ideal for: Camera, Lenses/Filters, Microscopes, Computer, Eyeglasses, Plastic, Metal. Machine or hand washable for long lasting use.
Catalog No. 50949540
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.