missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Luminata Forte ELISA HRP Substrate

Catalog No. elluf0100
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ELLUF0100 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ELLUF0100 Supplier MilliporeSigma™ Supplier No. ELLUF0100
Only null left

Used in Western blotting; dot/slot blotting

  • Detection Methods: Chemiluminescence
  • Key Applications: ELISA
  • Chemiluminescent: HRP detection reagent
  • For use on immunoblots

Specifications

Quantity 100 mL
Storage Requirements 2° to 8°C