missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mopec Lumea BxBoard Prostate Fusion Kit
Description
- Includes:
- 1 - 6-Site Left (Green) BxBoard (lat base, base, lat mid, mid, lat apex, apex)
- 1 - 6-Site Right (Blue) BxBoard (base, lat base, mid, lat mid, apex, lat apex)
- 1 - X1 Outlier (Black) BxBoard
- 1 - X2 Outlier (Black) BxBoard
- 1 - X3 Outlier (Black) BxBoard
- 1 - FUSION Biopsy Corrections/Additional Samples Form
- 1 - Protective Transport Mailer Box (6.5 x 5.5 x 3.75 in.)
- Hydrophilic formalin-soaked sponge technology preserves anatomic orientation of biopsy tissue.
- Designed to reduce formaldehyde exposure for medical assistants, physicians, and laboratory personnel.
- Clinics report an 83% reduction in specimen containers and an 89% reduction in formalin per prostate case.
- Provides an overall 136% reduction in biohazard waste per histology lab.
- Mopec is the exclusive supplier of Lumea™ consumable products.
Specifications
Specifications
| Sterility | Non-sterile |
| Color | Black |
| Tip Material | Sponge |
| Material | Polypropylene |