missing translation for 'onlineSavingsMsg'
Learn More

Carson™ Lume Series Aspheric COB LED Lighted Hand-Held Magnifier with 7x Spot Lens and Two Brightness Settings DFS Item

Magnifying glasses contain three ultra bright COB LED lights spread proportionally around rim of lens for wide and even lighting

Supplier:  Carson™ AS90

 View more versions of this product

Catalog No. S81724

Only null left
Explore available promotions

Description

Description

  • For low vision tasks including reading fine print, hobby, crafts, coin and stamp inspection, model building, jewelry making
  • Lightweight, acrylic lenses have aspheric design
    • Light rays pass through lens and meet at a single point for crystal clear and distortion-free image quality
  • Backed by Carson™'s limited lifetime warranty
  • COB LED lights powered by 2 AAA batteries (not included)
  • Magnifiers available in two different magnifications:
    • 2.5x Power (AS-90)
      • Stronger magnification but smaller lens diameter (3.5 in.)
    • 2x power (AS-95)
      • Provides a wider viewing area with 4 in. lens diameter
  • Includes:
    • 7x power spot lens built into handle at magnifier
    • Protective rim around lens to protect from scratching and breaking
Specifications

Specifications

0.089 m
Acrylic
2.5x
Magnifier, Handheld Lighted
3.5 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions