missing translation for 'onlineSavingsMsg'
Learn More

Lumazine 99.0+%, TCI America™

Catalog No. p-7131815
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L00861G 1 g
L00865G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L00861G Supplier TCI America Supplier No. L00861G
Only null left

Chemical Identifiers

CAS 487-21-8
Molecular Formula C6H4N4O2
Molecular Weight (g/mol) 164.12
MDL Number MFCD00066250
InChI Key UYEUUXMDVNYCAM-UHFFFAOYSA-N
Synonym lumazine, 2,4-pteridinediol, 2,4-dihydroxypteridine, 2,4 1h,3h-pteridinedione, pteridine-2,4-dione, lumazin, 2,4 3h,8h-pteridinedione, pteridine-2,4 1h,3h-dione, pteridine-2,4-diol, unii-260mf9u4wc
PubChem CID 10250
ChEBI CHEBI:16489
IUPAC Name 1,2,3,4-tetrahydropteridine-2,4-dione
SMILES O=C1NC(=O)C2=NC=CN=C2N1

Specifications

Quantity 1 g
Formula Weight 164.12
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Lumazine
RTECSNumber UO3416000
TSCA Yes