missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lumazine 99.0+%, TCI America™
Specifications
Specifications
| Quantity | 5 g |
| Formula Weight | 164.12 |
| Percent Purity | ≥99.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Lumazine |
Safety and Handling
| RTECSNumber | UO3416000 |
| TSCA | Yes |