Learn More
MilliporeSigma™ Lugol's solution (Diluted Iodine-Potassium Iodide Solution)
Catalog No. m1092611000
Change view
Click to view available options
Volume (Metric):
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
M1092611000 1 L
M1092612500 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1092611000 Supplier MilliporeSigma™ Supplier No. 1.09261.1000
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
For staining of Gram bacteria
  • Allows fast differentiation of bacteria in Gram-positive and Gram-negative
  • Mureine structure of bacteria walls is the basis of the color affinity
  • Bacteria will be stained with Gram's crystal violet solution, aniline dye
  • Gram-negative bacteria will be decolorized
  • Gram-negative bacteria will be counterstained by Gram's safranin solution, appear orange
Specifications
Volume (Metric) 1 L
Packaging Plastic Bottle