Learn More
MilliporeSigma™ Lugol's solution (Diluted Iodine-Potassium Iodide Solution)
For staining of Gram bacteria
$255.76 - $536.51
| Catalog Number | Mfr. No. | Volume (Metric) | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Volume (Metric) | Price | Quantity & Availability | |||||
|
M1092611000
|
MilliporeSigma™
1.09261.1000 |
1 L |
Each for $255.76
|
|
|||||
|
M1092612500
|
MilliporeSigma™
1.09261.2500 |
2.5 L |
Each for $536.51
|
|
|||||
Description
- Allows fast differentiation of bacteria in Gram-positive and Gram-negative
- Mureine structure of bacteria walls is the basis of the color affinity
- Bacteria will be stained with Gram's crystal violet solution, aniline dye
- Gram-negative bacteria will be decolorized
- Gram-negative bacteria will be counterstained by Gram's safranin solution, appear orange
Specifications
| Plastic Bottle |