Learn More
Air-Tite Luer Lock Syringe with Needle
Description
- EZ Dose Luer Lock Syringe W/ 25G X 5/8 in. Needle 100/Box
Specifications
Specifications
| Material | Polypropylene (Barrel), Polypropylene (Plunger), Polypropylene (Hub), Polyisoprene Rubber (Tip) |
| Needle Gauge | 25 G |
| Latex Free | Yes |
| Product Type | Luer Lock Syringe with Needle |
| Disposable | Single-use |
| Volume (Metric) | 1 mL |
| Connection Type | Centric Luer Lock |
| Graduated | Yes |
| Sterility | Sterile |
General Lab and Vet use only