LGC Genomics Llc (Lucigen) E. cloni 5-alpha Chemically Competent Cells, 12 Rxns (DUOs)Clone and subclone with high efficiency, chemically competent DH5-alpha cells.
Catalog No. NC2162032
Direct replacements for standard cloning strainsOptimized genetics for high yields: endonuclease and recombination minus, for blue/white screening-capable.Competent Cells include Control DNA and Recovery Medium, and are packaged as DUOs (2 transformations per tube). Recovery Medium is also available separately. The specified transformation efficiencies are with pUC DNA, unless indicated otherwise.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Direct replacements for standard cloning strainsOptimized genetics for high yields: endonuclease and recombination minus, for blue/white screening-capable.Competent Cells include Control DNA and Recovery Medium, and are packaged as DUOs (2 transformations per tube). Recovery Medium is also available separately. The specified transformation efficiencies are with pUC DNA, unless indicated otherwise.