missing translation for 'onlineSavingsMsg'
Learn More

Promega Luciferin Detection Reagent

A reagent to detect presence of pro-luciferin substrates used to monitor the activity of specific isoforms of cytochrome P450 or NAT2.

$255.61 - $612.68

Specifications

Content And Storage -30°C to -10°C
For Use With (Application) Monitor the activity of specific isoforms of cytochrome P450 or NAT2
pH 7.2
Product Type Luciferin
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PRV8920
Encompass
View Documents Promotion Details
Promega
V8920
10 mL
Each for $255.61
Only null left
 
PRV8921
Encompass
View Documents Promotion Details
Promega
V8921
50 mL
Each for $612.68
Only null left
 
Description

Description

  • Reagent for Detection of Pro-Luciferin Substrates
  • Reagent available without esterase
  • Use in combination with the pro-luciferin substrates
Specifications

Specifications

-30°C to -10°C
Monitor the activity of specific isoforms of cytochrome P450 or NAT2
7.2
Luciferin
Videos
SDS
Documents

Documents