missing translation for 'onlineSavingsMsg'
Learn More

Corning™ LSE™ Fixed-Angle Rotors

Catalog No. 07201418
Change view
Click to view available options
Capacity:
18 x 1.5 mL
6 x 50 mL
12 x 15 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity
07201418 6 x 50 mL
07201421 18 x 1.5 mL
07201419 12 x 15 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 07201418 Supplier Corning™ Supplier No. 480136

Rotors with different tube size and capacity configurations

This is an optional rotor for 6 x 50 mL tubes, for use with Corning™ LSETM Compact Centrifuge.

Specifications

Type Rotor
Capacity 6 x 50 mL
Max. Speed 6,000 rpm
For Use With (Equipment) Corning™ LSE™ Compact Centrifuge
Angle Fixed