Learn More
Beckman Coulter LS 13 320 XR Particle Size Analyzer Sonicator Kit
Description
- An optional accessory for use with LS 13 320 XR Universal Liquid Module
- Needle probe sonicator for additional dispersion control of wet samples
- In-situ sonication with ULM before/during the run, can also be operated external to module
- Fully adjustable power settings
Specifications
Specifications
| Product Type | Sonicator Kit |
| For Use With (Equipment) | LS 13 320 XR Particle Size Analyzer with ULM |