Learn More
FUJIFILM Wako LPS For Endotoxin Indicator
Catalog No. 12333055
Change view
Click to view available options
Quantity:
6 Vials
Catalog No. Quantity
12333055 6 Vials
1 options
Catalog No. 12333055 Supplier FUJIFILM Wako Supplier No. WLPS0100K
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Composed of E. coli O55:B5, serves as stock solution to prepare multiple endotoxin indicators for depyrogenation validation studies
  • E. coli O55:B5
  • Can make multiple endotoxin indicators from one vial
  • Used for validation studies
Specifications
Form Lyophilized
Fill Volume >100,000 EU
Includes 6 vials containing >100,000 EU/vial
Quantity 6 Vials