Learn More
STERIS LpH™ III se Phenolic Disinfectant
This Phenolic Disinfectant is the latest generation of low pH disinfectant and is specifically formulated for the routine disinfection of hard, non-porous surfaces.
Supplier:  STERIS 647008CA
 View more versions of this product
Catalog No. 17463009
Only null left
Description
Description
  • A liquid concentrate specifically formulated for the disinfection of hard, non-porous surfaces
  • Use-dilution of rate of 0.8% v/v (1:128), solution can be applied manually or sprayed
  • Phenolic active ingredients provide broad spectrum antimicrobial efficacy against common bacteria and fungi
  • Can be used alone or in rotation with the Vesphene™ III Disinfectants
  • Ideal for use in a contamination control rotation, formulation includes surfactants for surface cleaning while disinfecting
  • Products include an extensive documentation package to meet validation objectives
  • Supported by the STERIS Technical Services team for individualized application and validation assistance
  • Each lot documented by Certificate of Analysis
Specifications
Chemical Identifiers
Phenolic Disinfectant
Non-sterile
0.8% v/v (1:128)
Specifications
Phenolic Disinfectant
0.8% v/v (1:128)
4/Cs.
Non-sterile
Disinfecting Hard, Non-Porous Surfaces
1 gal.
Product Suggestions
Product Suggestions