missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ LPD-12, Calbiochem™,

Catalog No. 4376001MG
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4376001MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4376001MG Supplier MilliporeSigma™ Supplier No. 4376001MG

Specifications

Color White
Quantity 1 mg
Packaging Glass Bottle
Molecular Formula C131H224N32O37
Synonym Lipopeptide Detergent-12, Ac-AO(C12)AEAAEKAAKYAAEAAEKAAKAO(C12)A-NH2
Percent Purity ≥95% (HPLC)