missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Loxoprofen 97.0+%, TCI America™
Supplier: TCI America L02445G
Specifications
| Loxoprofen | |
| 111°C | |
| C15H18O3 | |
| 5 g | |
| YMBXTVYHTMGZDW-UHFFFAOYNA-N | |
| 2-{4-[(2-oxocyclopentyl)methyl]phenyl}propanoic acid | |
| 3965 | |
| 246.31 | |
| Crystalline Powder |
| 68767-14-6 | |
| White-Yellow | |
| MFCD00864331 | |
| 2-[4-[(2-Oxocyclopentyl)methyl]phenyl]propionic Acid | |
| CC(C(O)=O)C1=CC=C(CC2CCCC2=O)C=C1 | |
| 246.31 | |
| CHEBI:76172 | |
| ≥97.0% (HPLC,T) |
Chemical Identifiers
| 68767-14-6 | |
| 246.31 | |
| YMBXTVYHTMGZDW-UHFFFAOYNA-N | |
| 3965 | |
| 2-{4-[(2-oxocyclopentyl)methyl]phenyl}propanoic acid |
| C15H18O3 | |
| MFCD00864331 | |
| 2-[4-[(2-Oxocyclopentyl)methyl]phenyl]propionic Acid | |
| CHEBI:76172 | |
| CC(C(O)=O)C1=CC=C(CC2CCCC2=O)C=C1 |
Safety and Handling
TSCA : No