missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corepoint Scientific™ Lower Glycerol Bottle Kit
Description
- Lower glycerol bottle kit
- Designed for Corepoint Scientific™ Blood Bank Refrigerators and Plasma Freezers
Specifications
Specifications
| Style | Upright |
| Color | White |
| For Use With (Application) | Glycerol Kit to replace lower solid ballast |
| For Use With (Equipment) | Corepoint™ Blood Bank Refrigerators and Plasma Freezers |
| Model | NSBBP GLYCEROL KIT |