Learn More
Thermo Scientific™ Lowenstein-Jensen Medium (LJ)
Reduce media preparation time and simultaneously cultivate and isolate Mycobacterium spp. using convenient, pre-filled, ready-to-use tubes.
$761.74 - $1045.96
Specifications
Manufacturing Origin United States
Media Type Lowenstein-Jensen Medium (LJ)
Product Type Microbiology Media
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Format Media Type Manufacturing Origin Price Quantity  
Catalog Number Mfr. No. Quantity Format Media Type Manufacturing Origin Price Quantity  
R08500
View Documents
Thermo Scientific™
RE08500
100/Pk. Tube Lowenstein-Jensen Medium (LJ) United States
Case of 100 Each for $1,045.96
Only null left
R08502
View Documents
Thermo Scientific™
RE08502
20/Pk. Tube Lowenstein-Jensen Medium (LJ) United States
Pack of 20 Each for $761.74
Only null left
Description
Description
Cultivate and isolate Mycobacterium spp. using Thermo Scientific™ Lowenstein-Jensen Medium (LJ).
Remove the need for pipetting, dispensing or autoclaving with convenient, pre-filled and ready-to-use tubes and bottles of Lowenstein-Jensen Medium, a glycerated egg-potato medium which provides fatty acids and protein required for metabolsm of mycobacteria. Malachite green is added to inhibit other mycobacteria and as a pH indicator.
Thermo Scientific Lowenstein-Jensen Medium (LJ) agar slants are available in a variety of fill volumes and pack sizes.
Mycobacteria most commonly involve infections of the pulmonary system, but some species infect the lymphatic system and skin. Mycobacteria are gram-positive, nonmotile, acid-fast pleomorphic rods. They vary in colony morphology, pigmentation, and growth rates. Mycobacterium tuberculosis, the most important pathogenic Mycobacterium, is slow-growing and causes highly contageous, airborne respiratory disease. Tuberculosis is responsible for an estimated 2 million deaths per year worldwide.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Specifications
United States
Lowenstein-Jensen Medium (LJ)
Microbiology Media
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'