missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Lowenstein-Jensen Medium (LJ)

Catalog No. R08500
Change view
Click to view available options
Quantity:
100/Pk.
20/Pk.
Format:
Tube
Manufacturing Origin:
United States
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format Manufacturing Origin
R08500 100/Pk. Tube United States
R08502 20/Pk. Tube United States
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R08500 Supplier Thermo Scientific™ Supplier No. RE08500

Reduce media preparation time and simultaneously cultivate and isolate Mycobacterium spp. using convenient, pre-filled, ready-to-use tubes.

Cultivate and isolate Mycobacterium spp. using Thermo Scientific™ Lowenstein-Jensen Medium (LJ).

Remove the need for pipetting, dispensing or autoclaving with convenient, pre-filled and ready-to-use tubes and bottles of Lowenstein-Jensen Medium, a glycerated egg-potato medium which provides fatty acids and protein required for metabolsm of mycobacteria. Malachite green is added to inhibit other mycobacteria and as a pH indicator.

Thermo Scientific Lowenstein-Jensen Medium (LJ) agar slants are available in a variety of fill volumes and pack sizes.

Mycobacteria most commonly involve infections of the pulmonary system, but some species infect the lymphatic system and skin. Mycobacteria are gram-positive, nonmotile, acid-fast pleomorphic rods. They vary in colony morphology, pigmentation, and growth rates. Mycobacterium tuberculosis, the most important pathogenic Mycobacterium, is slow-growing and causes highly contageous, airborne respiratory disease. Tuberculosis is responsible for an estimated 2 million deaths per year worldwide.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

Specifications

Color Opaque agar, pale green
Content And Storage 2°C to 8°C
Format Tube
Manufacturing Origin United States
Media Type Lowenstein-Jensen Medium (LJ)
Product Type Prepared Microbiology Tubes
Packaging Type Glass tube with black screw cap
Quantity 100/Pk.