missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Low Profile Tubes, strips of 8, white

Catalog No. AB0771W
Change view
Click to view available options
Quantity:
120 strips
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB0771W 120 strips
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB0771W Supplier Thermo Scientific™ Supplier No. AB0771W

Increase the efficiency of reactions with these low-profile strip tubes for PCR and qPCR applications.

The low profile design of the Thermo Scientific strip tubes without caps reduces the 'dead space' between the heated lid of the thermal cycler and the sample. This eliminates condensation on the side wall of the tube, preventing reduction in PCR volume and increasing the efficiency of the reaction. Low profile products are especially recommended for use with reaction volumes below 20 μL.

Highlights

  • Suitable for 0.2 mL thermal cycler blocks
  • 150 µL maximum tube capacity when capped
  • Low profile to reduce dead space and increase PCR efficiency
  • Labelled A-H end tabs (8-strip) and 1-12 end tabs (12-strip)
  • Sealable with Flat, Domed, and Ultra Clear Cap Strips

Specifications

Certifications/Compliance Certified DNA/RNase Free
Color White
Volume (Metric) Working 0.1 mL
Volume (Metric) 0.15 mL
Quantity 120 strips
For Use With (Equipment) Thermal Cyclers
Sterility Non-sterile
Material Polypropylene
Reaction Speed Low profile
Closure Type Not Included
Format 120 Strip Tubes
Product Type 8-Strip
Show More Show Less