Learn More
Idex Low-Pressure PEEK Tees
Combine, split, or intersect fluid streams throughout your system
Supplier: Idex UPP 713
Description
- Manufactured from inert polymers
- 1/4-28 flat-bottom ports
Use Tee connectors in a variety of low-pressure situations throughout your fluidic system. Handy mounting holes make it a snap to mount connectors. Tee connector assemblies come complete with nuts and ferrules.
Chemical Identifiers
| 1000 psi | |
| PEEK |
Specifications
| Union | |
| 1000 psi | |
| 1.25 mm |
| 0.050 in. | |
| PEEK |